Learn More
Biomatik Corporation Mouse Secreted Frizzled Related Protein 4 (SFRP4) ELISA Kit
Supplier:  Biomatik Corporation EKU07220
Encompass_Preferred
Mouse Secreted Frizzled Related Protein 4 (SFRP4) ELISA Kit for detecting SFRP4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z1N6. Gene ID: 20379. Target Synonyms: FRP-4; FRPHE; Frizzled protein, human endometrium. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2860