Learn More
Biomatik Corporation Human Sialoadhesin (SN) ELISA Kit
Supplier:  Biomatik Corporation EKU07325
Encompass_Preferred
Human Sialoadhesin (SN) ELISA Kit for detecting SN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BZZ2. Gene ID: 6614. Target Synonyms: CD169; SIGLEC1; Sialic Acid Binding Ig Like Lectin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2877