Learn More
Biomatik Corporation Human Sirtuin 5 (SIRT5) ELISA Kit
Supplier:  Biomatik Corporation EKU07356
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.062ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9NXA8. Target Synonyms: SIR2L5; Silent Mating Type Information Regulation 2 Homolog 5; NAD-dependent protein deacylase sirtuin-5, mitochondrial; Regulatory protein SIR2 homolog 5; SIR2-like protein 5. Restriction: For Research Use Only.
Catalog No. 50-229-2882