Learn More
Biomatik Corporation Human Sodium/Hydrogen Exchanger 1 (NHE1) ELISA Kit
Supplier:  Biomatik Corporation EKU07388
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: SLC9A1; APNH; Solute Carrier Family 9 Member 1,Sodium/Hydrogen Exchanger; Antiporter,Na+/H+,Amiloride Sensitive; Na(+)/H(+) antiporter, amiloride-sensitive. Restriction: For Research Use Only.
Catalog No. 50-229-2887