Learn More
Biomatik Corporation Human Solute Carrier Family 39, Member 6 (SLC39A6) ELISA Kit
Supplier:  Biomatik Corporation EKU07407
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q13433. Target Synonyms: LIV-1; Zinc Transporter Protein 6; Estrogen-regulated protein LIV-1; Zrt- and Irt-like protein 6; Zinc transporter ZIP6. Restriction: For Research Use Only.
Catalog No. 50-229-2890