Learn More
Biomatik Corporation Human Spectrin Beta, Non Erythrocytic 4 (SPTbN4) ELISA Kit
Supplier:  Biomatik Corporation EKU07423
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.063ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9H254. Target Synonyms: QV; SPNB4; SPTBN3; Beta-IV spectrin; Spectrin, non-erythroid beta chain 3. Restriction: For Research Use Only.
Catalog No. 50-229-2893