Learn More
Biomatik Corporation Rat Steroid 5 Alpha Reductase 1 (SRD5a1) ELISA Kit
Supplier:  Biomatik Corporation EKU07470
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.056ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P24008. Target Synonyms: 3-Oxo-5 Alpha-Steroid Delta 4-Dehydrogenase Alpha 1. Restriction: For Research Use Only.
Catalog No. 50-229-2899