Learn More
Biomatik Corporation Mouse Succinate Dehydrogenase Complex Subunit D (SDHD) ELISA Kit
Supplier:  Biomatik Corporation EKU07490
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9CXV1. Target Synonyms: PGL; PGL1; QPs3; Succinate-ubiquinone reductase membrane anchor subunit; Succinate dehydrogenase [ubiquinone] cytochrome b small subunit, mitochondrial. Restriction: For Research Use Only.
Catalog No. 50-229-2902