Learn More
Biomatik Corporation Human Superoxide Dismutase Copper Chaperone ELISA Kit
Supplier:  Biomatik Corporation EKU07508
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O14618. Target Synonyms: CCS; Copper Chaperone For Superoxide Dismutase. Restriction: For Research Use Only.
Catalog No. 50-229-2907