Learn More
Biomatik Corporation Rat Surfactant Protein C (SP-C) ELISA Kit
Supplier:  Biomatik Corporation EKU07528
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, lung lavage fluid and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.112ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P11685. Target Synonyms: SFTPC; PSP-C; SFTP2; SPC; SP5; Surfactant Associated Protein C; Pulmonary Surfactant Protein C; Pulmonary surfactant-associated proteolipid SPL(Val). Restriction: For Research Use Only.
Catalog No. 50-229-2912