Learn More
Biomatik Corporation Human Talin 1 (TLN1) ELISA Kit
Supplier:  Biomatik Corporation EKU07566
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9Y490. Target Synonyms: Talin Protein; TLN; ILWEQ. Restriction: For Research Use Only.
Catalog No. 50-229-2915