Learn More
Biomatik Corporation Human Teratocarcinoma Derived Growth Factor 1 (TDGF1) ELISA Kit
Supplier:  Biomatik Corporation EKU07597
Encompass_Preferred
Human Teratocarcinoma Derived Growth Factor 1 (TDGF1) ELISA Kit for detecting TDGF1 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13385. Gene ID: 6997. Target Synonyms: CR; CRGF; CRIPTO; Cripto-1; Epidermal growth factor-like cripto protein CR1; Cripto-1 growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2923