Learn More
Biomatik Corporation Human Thimet Oligopeptidase 1 (THOP1) ELISA Kit
Supplier:  Biomatik Corporation EKU07611
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 1.56-100ng/mL. Sensitivity: 0.63ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P52888. Target Synonyms: EP24.15; MEPD_HUMAN; MP78; Endopeptidase 24.15. Restriction: For Research Use Only.
Catalog No. 50-229-2925