Learn More
Biomatik Corporation Bovine Thrombospondin 1 (THBS1) ELISA Kit
Supplier:  Biomatik Corporation EKU07639
Encompass_Preferred
Bovine Thrombospondin 1 (THBS1) ELISA Kit for detecting THBS1 in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 78-5,000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28178. Gene ID: 281530. Target Synonyms: TSP1; Thrombospondin-1p180. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2929