Learn More
Biomatik Corporation Rat Thyroglobulin (TG) ELISA Kit
Supplier:  Biomatik Corporation EKU07674
Encompass_Preferred
Rat Thyroglobulin (TG) ELISA Kit for detecting TG in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06882. Target Synonyms: AITD3; TGN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2935