Learn More
Biomatik Corporation Rat Tight Junction Protein 1 (TJP1) ELISA Kit
Supplier:  Biomatik Corporation EKU07694
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.056ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: F1M4A0. Target Synonyms: ZO1; Zona Occludens 1; Zona occludens protein 1; Zonula occludens protein 1. Restriction: For Research Use Only.
Catalog No. 50-229-2939