Learn More
Biomatik Corporation Pig Tissue Inhibitors Of Metalloproteinase 2 (TIMP2) ELISA Kit
Supplier:  Biomatik Corporation EKU07718
Encompass_Preferred
Pig Tissue Inhibitors Of Metalloproteinase 2 (TIMP2) ELISA Kit for detecting TIMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: C0JPM4. Gene ID: 396988. Target Synonyms: CSC-21K; TIMP Metallopeptidase Inhibitor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2942