Learn More
Biomatik Corporation Mouse Toll Like Receptor 3 (TLR3) ELISA Kit
Supplier:  Biomatik Corporation EKU07751
Encompass_Preferred
Mouse Toll Like Receptor 3 (TLR3) ELISA Kit for detecting TLR3 in saliva, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99MB1. Gene ID: 142980. Target Synonyms: CD283. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2949