Learn More
Biomatik Corporation Human Transforming Growth Factor Beta Receptor II (TGFbR2) ELISA Kit
Supplier:  Biomatik Corporation EKU07823
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.118ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P37173. Target Synonyms: MFS2. Restriction: For Research Use Only.
Catalog No. 50-229-2959