Learn More
Biomatik Corporation Human Tryptophan Hydroxylase 2 (TPH2) ELISA Kit
Supplier:  Biomatik Corporation EKU07930
Encompass_Preferred
Human Tryptophan Hydroxylase 2 (TPH2) ELISA Kit for detecting TPH2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8IWU9. Gene ID: 121278. Target Synonyms: NTPH; Neuronal tryptophan hydroxylase; Tryptophan 5-monooxygenase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2971