Learn More
Biomatik Corporation Mouse Vesicular Glutamate Transporter 1 (VGLUT1) ELISA Kit
Supplier:  Biomatik Corporation EKU08133
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: SLC17A7; BNPI; Solute Carrier Family 17 Member 7,Sodium-Dependent Inorganic Phosphate Cotransporter; Brain-specific Na(+)-dependent inorganic phosphate cotransporter. Restriction: For Research Use Only.
Catalog No. 50-229-2998