Learn More
Biomatik Corporation Rat Vitamin D Receptor (VDR) ELISA Kit
Supplier:  Biomatik Corporation EKU08169
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.625-40ng/mL. Sensitivity: 0.239ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: NR1I1; Nuclear Receptor Subfamily 1 Group I Member 1; Calcitriol Receptor; 1,25-dihydroxyvitamin D3 receptor. Restriction: For Research Use Only.
Catalog No. 50-229-3005