Learn More
Biomatik Corporation Human V-Yes-1 Yamaguchi Sarcoma Viral Related Oncogene Homolog (LYN) ELISA Kit
Supplier:  Biomatik Corporation EKU08197
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.062ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P07948. Target Synonyms: JTK8; p53Lyn; p56Lyn; Tyrosine-protein kinase Lyn; Lck/Yes-related novel protein tyrosine kinase. Restriction: For Research Use Only.
Catalog No. 50-229-3009