Learn More
Biomatik Corporation Human Thrombospondin 3 (THBS3) ELISA Kit
Supplier:  Biomatik Corporation EKU08294
Encompass_Preferred
Human Thrombospondin 3 (THBS3) ELISA Kit for detecting THBS3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49746. Gene ID: 7059. Target Synonyms: TSP3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3018