Learn More
Biomatik Corporation Mouse Pentraxin 3, Long (PTX3) ELISA Kit
Supplier:  Biomatik Corporation EKU08315
Encompass_Preferred
Mouse Pentraxin 3, Long (PTX3) ELISA Kit for detecting PTX3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48759. Gene ID: 19288. Target Synonyms: TNFAIP5; TSG-14; Pentaxin-Related Gene,Rapidly Induced By IL-1 Beta; Tumor Necrosis Factor Alpha-Induced Protein 5; Tumor necrosis factor-inducible gene 14 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3022