Learn More
Biomatik Corporation Human Insulin Receptor Substrate 1 (IRS1) ELISA Kit
Supplier:  Biomatik Corporation EKU08337
Encompass_Preferred
Human Insulin Receptor Substrate 1 (IRS1) ELISA Kit for detecting IRS1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35568. Gene ID: 3667. Target Synonyms: HIRS1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3026