Learn More
Biomatik Corporation Mouse Crystallin Beta A1 (CRYbA1) ELISA Kit
Supplier:  Biomatik Corporation EKU08343
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.118ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P02525. Target Synonyms: CRYB1; Eye Lens Structural Protein. Restriction: For Research Use Only.
Catalog No. 50-229-3028