Learn More
Biomatik Corporation Rat Uroplakin 3A (UPK3A) ELISA Kit
Supplier:  Biomatik Corporation EKU08377
Encompass_Preferred
Rat Uroplakin 3A (UPK3A) ELISA Kit for detecting UPK3A in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZZ76. Gene ID: 315190. Target Synonyms: UPIII; UPK3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3035