Learn More
Biomatik Corporation Human Adenylyl Cyclase Associated Protein 1 (CAP1) ELISA Kit
Supplier:  Biomatik Corporation EKU08477
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.064ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q01518. Target Synonyms: CAP; CAP1-PEN. Restriction: For Research Use Only.
Catalog No. 50-229-3052