Learn More
Biomatik Corporation Mouse Meprin A Alpha (MEP1a) ELISA Kit
Supplier:  Biomatik Corporation EKU08670
Encompass_Preferred
Mouse Meprin A Alpha (MEP1a) ELISA Kit for detecting MEP1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 49.4-4,000pg/ml. Sensitivity: 19.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P28825. Gene ID: 17287. Target Synonyms: PPHA; PABA Peptide Hydrolase; Endopeptidase-2; N-benzoyl-L-tyrosyl-P-amino-Benzoic Acid Hydrolase Subunit Alpha; PABA Peptide Hydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3057