Learn More
Biomatik Corporation Bovine Follicle Stimulating Hormone (FSH) ELISA Kit
Supplier:  Biomatik Corporation EKU08703
Encompass_Preferred
Bovine Follicle Stimulating Hormone (FSH) ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.91ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01217; P04837. Gene ID: 280749; 281171. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3060