Learn More
Biomatik Corporation Bovine Adrenocorticotropic Hormone (ACTH) ELISA Kit
Supplier:  Biomatik Corporation EKU08709
Encompass_Preferred
Bovine Adrenocorticotropic Hormone (ACTH) ELISA Kit for detecting ACTH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 12.35-1000pg/ml. Sensitivity: 4.57pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01190. Gene ID: 281416. Target Synonyms: Corticotropin; Adrenocorticotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3062