Learn More
Biomatik Corporation Rat Cathelicidin Antimicrobial Peptide (CAMP) ELISA Kit
Supplier:  Biomatik Corporation EKU08738
Encompass_Preferred
Rat Cathelicidin Antimicrobial Peptide (CAMP) ELISA Kit for detecting CAMP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 24.69-2000ng/ml. Sensitivity: 9.16ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: G3V8S9. Gene ID: 316010. Target Synonyms: CAP18; FALL39; HSD26; LL37; 18 kDa Cationic Antimicrobial Protein; Antibacterial protein FALL-39; FALL-39 peptide antibiotic. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3064