Learn More
Biomatik Corporation Rabbit Aspartate Aminotransferase (AST) ELISA Kit
Supplier:  Biomatik Corporation EKU09053
Encompass_Preferred
Sample Type: Serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Rabbit. Detection Range: 0.625-40ng/mL. Sensitivity: 0.299ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: cCAT; AAT; ASAT; SGOT; GOT1; Cysteine transaminase, cytoplasmic; Transaminase A; Aspartate Transaminase 1,Cytoplasmic; Glutamic-Oxaloacetic Transaminase 1,Soluble. Restriction: For Research Use Only.
Catalog No. 50-229-3086