Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily, Member 9 (TNFRSF9) ELISA Kit
Supplier:  Biomatik Corporation EKU09067
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily, Member 9 (TNFRSF9) ELISA Kit for detecting TNFRSF9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07011. Target Synonyms: CD137; CDw137; 4-1BB; ILA; Induced By Lymphocyte Activation; T-cell antigen 4-1BB homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3088