Learn More
Biomatik Corporation Human/Mouse/Rat Lamin B1 (LMNB1) ELISA Kit
Supplier:  Biomatik Corporation EKU09218
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human/Mouse/Rat. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: LMN; LMN2; LMNB. Restriction: For Research Use Only.
Catalog No. 50-229-3102