Learn More
Biomatik Corporation Human Inositol Polyphosphate-5-Phosphatase 145kDa (INPP5D) ELISA Kit
Supplier:  Biomatik Corporation EKU09324
Encompass_Preferred
Human Inositol Polyphosphate-5-Phosphatase 145kDa (INPP5D) ELISA Kit for detecting INPP5D in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92835. Gene ID: 3635. Target Synonyms: SHIP; SHIP1; SIP-145; hp51CN; Phosphatidylinositol-3,4,5-Trisphosphate 5-Phosphatase 1; SH2 domain-containing inositol 5'-phosphatase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3117