Learn More
Biomatik Corporation Human Adrenocorticotropic Hormone (ACTH) CLIA Kit
Supplier:  Biomatik Corporation EKU09474
Encompass_Preferred
Human Adrenocorticotropic Hormone (ACTH) CLIA Kit for detecting ACTH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.9-1,000pg/ml. Sensitivity: 1.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P17858. Gene ID: 5211. Target Synonyms: Corticotropin; Adrenocorticotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3133