Learn More
Biomatik Corporation Human Interleukin 1 Zeta (IL1z) CLIA Kit
Supplier:  Biomatik Corporation EKU09567
Encompass_Preferred
Human Interleukin 1 Zeta (IL1z) CLIA Kit for detecting IL1z in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.51pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9NZH6. Gene ID: 27178. Target Synonyms: IL1F7; IL37; FIL1; FIL1 (ZETA) ; FIL1Z; IL-1F7; IL-1H4; IL-1RP1; IL1H4; IL1RP1; Interleukin 37; Interleukin-1-Related Protein; Interleukin 1 Family, Member 7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3137