Learn More
Biomatik Corporation Rat Interferon Alpha 13 (IFNa13) CLIA Kit
Supplier:  Biomatik Corporation EKU09646
Encompass_Preferred
Rat Interferon Alpha 13 (IFNa13) CLIA Kit for detecting IFNa13 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.52pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P05011. Gene ID: 298210. Target Synonyms: LeIF D; Interferon alpha-D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3139