Learn More
Biomatik Corporation Rat Colony Stimulating Factor 3, Granulocyte (GCSF) CLIA Kit
Supplier:  Biomatik Corporation EKU09752
Encompass_Preferred
Rat Colony Stimulating Factor 3, Granulocyte (GCSF) CLIA Kit for detecting GCSF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.69-500pg/ml. Sensitivity: 0.29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P97712. Target Synonyms: CSF3; G-CSF; Granulocyte Colony Stimulating Factor; Pluripoietin; Filgrastim; Lenograstim. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3141