Learn More
Biomatik Corporation Rat Adrenomedullin (ADM) ELISA Kit
Supplier:  Biomatik Corporation EKU10058
Encompass_Preferred
Rat Adrenomedullin (ADM) ELISA Kit for detecting ADM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P43145. Gene ID: 25026. Target Synonyms: AM; PAMP; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3152