Learn More
Biomatik Corporation Human Cluster Of Differentiation 38 (CD38) ELISA Kit
Supplier:  Biomatik Corporation EKU10098
Encompass_Preferred
Human Cluster Of Differentiation 38 (CD38) ELISA Kit for detecting CD38 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.65ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28907. Gene ID: 952. Target Synonyms: cADPRH; ADPRC 1; c-ADPRH; T10; p45; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; ADP-ribosyl cyclase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3155