Learn More
Biomatik Corporation Mouse Neurofilament, Light Polypeptide (NEFL) ELISA Kit
Supplier:  Biomatik Corporation EKU10166
Encompass_Preferred
Mouse Neurofilament, Light Polypeptide (NEFL) ELISA Kit for detecting NEFL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 48.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P08551. Gene ID: 18039. Target Synonyms: CMT1F; CMT2E; NF-L; NF68; NFL; 68 kDa neurofilament protein; Neurofilament triplet L protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3160