Learn More
Biomatik Corporation Mouse Phenylalanine Hydroxylase (PAH) ELISA Kit
Supplier:  Biomatik Corporation EKU10195
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: PKU; PKU1; Phe-4-monooxygenase. Restriction: For Research Use Only.
Catalog No. 50-229-3164