Learn More
Biomatik Corporation Bovine C Reactive Protein (CRP) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10273
Encompass_Preferred
Bovine C Reactive Protein (CRP) High-sensitive ELISA Kit for detecting CRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: C4T8B4. Gene ID: 527553. Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3174