Learn More
Biomatik Corporation Human Interleukin 28B (IL28B) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10289
Encompass_Preferred
Human Interleukin 28B (IL28B) High-sensitive ELISA Kit for detecting IL28B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100pg/ml. Sensitivity: 0.64pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8IZI9. Gene ID: 282617. Target Synonyms: IFNL3; Interferon,Lambda 3; Cytokine Zcyto22. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3179