Learn More
Biomatik Corporation Rat CREB Binding Protein (CREBBP) ELISA Kit
Supplier:  Biomatik Corporation EKU10753
Encompass_Preferred
Rat CREB Binding Protein (CREBBP) ELISA Kit for detecting CREBBP in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6JHU9. Target Synonyms: CBP; RSTS; RTS; KAT3A; Rubinstein-Taybi Syndrome. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3215