Learn More
Biomatik Corporation Rabbit Cystatin C (Cys-C) ELISA Kit
Supplier:  Biomatik Corporation EKU10771
Encompass_Preferred
Rabbit Cystatin C (Cys-C) ELISA Kit for detecting Cys-C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O97862. Gene ID: 100009450. Target Synonyms: CST3; Cystatin 3; Gamma Trace; Post-Gamma-Globulin; Neuroendocrine Basic Polypeptide; Amyloid Angiopathy And Cerebral Hemorrhage. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3217