Learn More
Biomatik Corporation Rabbit Follicle Stimulating Hormone (FSH) ELISA Kit
Supplier:  Biomatik Corporation EKU10843
Encompass_Preferred
Rabbit Follicle Stimulating Hormone (FSH) ELISA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.85ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P07474; Q6IY73. Gene ID: 100009481; 100008946. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3230